Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 96 Well Round (U) Bottom Plate, TC Surface, Pack of 1
GSA_VA

Catalog No. 1256565 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
Case of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12-565-65 Case of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 12-565-65 Supplier Thermo Scientific™ Supplier No. 163320
Get 37% Off Cell Culture Plastics & More
Use promo code NUNC37
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal in a wide variety of applications, including cloning experiments and tissue culture-based analytical systems

Thermo Scientific™ Nunc MicroWell Plates with Nunclon Delta Surface are ideal in a wide variety of applications, including cloning experiments and tissue culture-based analytical systems. Thermo Scientific Nunc Cell Culture products offer solutions for cell growth and analysis and have been used by researchers worldwide since 1953. With a wide range of surface areas and treatments, we have products for all your basic or advanced cell-based research.

  • Tight-fitting lids reduce evaporation
  • Stackable
  • Radiation sterilized
  • Condensation rings control evaporation and retain condensate
  • Keyed lids prevent accidental reversals
  • Dimensions: 86 × 128mm

Ordering Information:

Individually wrapped.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Certified Nunclon Delta certified for monolayer formation, cloning efficiency, non-cytotoxic, non-pyrogenic, and sterility
Lid With Lid
Array 8 x 12
Packaging 1/Pk., 50/Cs.
Product Line Nunc
Color Clear
Product Type Microplate
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Nunclon Delta
Well Shape Round Bottom (U-well)
No. per Case 50
No. per Pack 1
Quantity Case of 50
Show More Show Less
What is the difference between the Nunclon Delta surface and a tissue culture (TC) surface from a competitor?

Our Nunclon Delta surface for adherent cells is a standard tissue culture (TC) surface modification that makes the polystyrene surface more hydrophilic. This facilitates maximum adhesion for a broad range of cell types.

The Nunclon Delta surface is Thermo Scientific Nunclon Delta-certified, which is a Thermo Fisher proprietary certification that has a quality standard to help ensure consistency. Every lot of Thermo Scientific Nunclon Delta-certified cell cultureware is tested with Gibco media for:
- Less than 10% growth variation in L929, Hel299 and primary chick embryo cells
- Less than 15% variation colony formation of V79-4 cells

More information and all applicable products can be found in the "Product selection guide for every step of your cell culture workflow" brochure .

What reagents are used to coat your Nunclon Delta surface products?

The Nunclon Delta surface is a surface modification that enhances cell attachment and growth for adherent cell lines and is not coated with any chemical reagent.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.