Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ Multidishes with Nunclon™ Supra Surface

Catalog No. p-200007434 Shop All Thermo Scientific Products
Change view
Click to view available options
No. of Wells:
6
12
24
48
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells
12-565-675 6
12-565-674 12
12-565-673 24
12-565-672 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12-565-675 Supplier Thermo Scientific™ Supplier No. 140680
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Experience optimal performance with Thermo Scientific Nunc plates with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells.

Experience optimal performance with Thermo Scientific Nunc plates with Nunclon Supra surface treatment for culturing a wide variety of fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.

Thermo Scientific Nunc vessels with Nunclon Supra surface treatment are designed for culturing fastidious adherent cells. Intended for general cell culture applications, the Nunclon Supra surface is more hydrophilic than the Nunclon Delta surface, enhancing the growth of finicky cell types under no-serum or low-serum conditions.

The Nunclon Supra surface supports serum-free and coating-free culture of human mesenchymal stromal cells (MSCs), retaining their morphology and promoting high cell viability. Nunclon Supra cultureware is a great xeno-free solution for cell therapy research applications.

Features of Nunc Multidishes with Nunclon Supra Surface include:
• Consistent lot-to-lot performance
• Sterile with SAL 10-6
• Non-cytotoxic
• Non-pyrogenic
• Biocompatible per USP Class VI for raw materials
• Shelf life: 5 years

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Lid With Lid
Array 2 x 3
Product Line Nunc
Color Clear
Disposable Single-use
Product Type Cell Culture Plate
Material Polystyrene
No. of Wells 6
Sterility Sterile
Surface Treatment Nunclon Supra
Well Shape Flat Bottom (F-well)
No. per Case 75 Dishes
No. per Pack 1 Dish
Show More Show Less

For Laboratory Use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.