Learn More
Thermo Scientific™ Nunc™ Multidishes with UpCell™ Surface
Catalog No. 03150019
Change view
Click to view available options
No. of Wells:
6
12
24
48
Culture Area:
1.1 sq. cm
1.9 sq. cm
3.5 sq. cm
9.6 sq. cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells Culture Area
03150019 48 1.1 sq. cm
03150020 24 1.9 sq. cm
03150021 12 3.5 sq. cm
03150022 6 9.6 sq. cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 03150019 Supplier Thermo Scientific™ Supplier No. 174898
Save 50% on Cell Culture Plastics
Use promo code NUNC50
View all promotions
Add to Cart
Add to Cart
Support non-enzymatic harvesting of adherent cells for preservation of cell viability and surface proteins.
Support non-enzymatic harvesting of adherent cells for preservation of cell viability and surface proteins with the temperature-responsive surface of Thermo Scientific™ Nunc™ UpCell™ Dishes. Simply transfer the culture to room temperature to collect cells in suspension or in the form of cell sheet with the help of a supportive membrane.
Traditional cell harvesting by enzymatic and mechanical methods often compromises the integrity of surface proteins and the viability of harvested cells. By contrast, UpCell Surface allows cell harvesting by simply reducing the temperature of the cell culture to below 32°C without the need of enzyme, resulting in highly viable cells with intact cell surface proteins.
  • No trypsinization—preserve cell surface proteins
  • No physical force—get high cell viability
  • For culture passaging, single-cell analyses and cell transplantation research
  • 6-well multidish comes with supportive membranes that enable harvesting of cell sheets and creations of 3-D tissue models
  • Minimal hands-on time
  • Customized bar coding is available
Recommended for:
Culture passaging; Single-cell analyses, Cell transplantation research, 3D tissue models
TRUSTED_SUSTAINABILITY
Specifications
Description 48 well, UpCell
Culture Area 1.1 sq. cm
Volume (Metric) Well 0.5 mL (Well)
Volume (Metric) Working 0.5 mL
Type Multi Dish
No. per Pack 1
No. per Case 6
Color Clear
Surface Treatment UpCell, Temperature Responsive
Sterility Sterile
No. of Wells 48
Well Shape Rectangular, Flat Bottom (F-well)
Show More Show Less
Why does temperature reduction detach cells from the Nunc dishes with UpCell Surface?
The UpCell Surface is composed of a covalently bound polymer that changes from being slightly hydrophobic to being very hydrophilic when the temperature is reduced to less than 32 degrees C. This results in the binding of water to the surface, swelling of the surface, and finally the release of the cells.
In cultureware with UpCell surface, does the temperature-responsive polymer detach with the cells?
The temperature-responsive polymer is covalently bound to the polystyrene dish or plate and will not detach from the product under normal cell culture procedures and conditions. The products have successfully passed the USP biological reactivity class VI testing.
What is special about the "UpCell surface with grid"?
The “UpCell surface with grid” products have a 3 x 3 mm grid design over the entire culture surface. Cell attachment and growth is restricted to within these small squares, preventing cells from clumping, and enabling easier harvest of the cells as single cells or small cell clusters. Products with “UpCell surface with grid” are suitable for the preparation of cells for flow cytometry analysis but cannot be used for cell-sheet applications. We offer 60 mm and 100 mm dishes with “UpCell surface with grid” whereas the 35 mm dishes contain supportive membrane for harvesting of cell sheets.
Which cell types can I use with the UpCell surface?
You can use the UpCell Surface with many cell types, including fibroblasts, mesenchymal stem cells, myocytes, keratinocytes, macrophages, and different epithelium and tumor cells. If cells do not detach spontaneously from the UpCell surface by lowering of the temperature to below 32 degrees C, the cells can be detached by flushing the surface with medium or by tapping on the side of the dish or plate. Cells that produce a lot of extracellular matrix and attach very strongly to the UpCell surface may be difficult to detach by temperature reduction alone. For these cell types, the simultaneous use of diluted trypsin is recommended.
Can I coat the UpCell surface with biomolecules to enable cell attachment?
Coating of the UpCell surface with biomolecules, for example, collagen, fibronectin, poly-lysine and laminin, is possible and should be done if you experience difficulties in attaching your cells to the UpCell surface. The coating procedure should be performed at 37 degrees C.
What should I do when cells cannot be collected as a cell sheet from the UpCell surface even if the supporting membrane is used?
The protocol for the detachment of cell sheets may need to be optimized for the cell type that you are using, particularly the degree of confluence at harvest, and the detachment conditions (temperature and incubation times).
Do I have to use a supporting membrane when collecting a cell sheet from the UpCell surface?
It is essential to use a supporting membrane when collecting the cells as a cell sheet from the UpCell Surface. However, you can use a hygroscopic PVDF membrane as an alternative to the supporting membrane supplied with the product (Cat. Nos. 174901 and 174904). Please note that the optimal protocol may vary, depending on the type of membrane used.
Can I use any temperature below 32 degrees C for detachment of cells from the UpCell surface?
Yes, the hydrophilicity of the UpCell Surface is constant below 32 degrees C. We recommend performing the detachment in a temperature-controlled, sterile environment. The detachment time would depend on the time it takes for the culture to be cooled down to less than 32 degrees C, as well as the cell type used.
Do cells detach from the UpCell surface during normal cell culture handling?
Detachment of cells from the UpCell surface occurs when the temperature of the culture is reduced below 32 degrees C. To avoid undesired cell detachment during handling, the following should be kept in mind:
- Use pre-warmed medium when changing the culture medium
- Observations under a microscope should be kept short to prevent cooling
How robust is the Nunc UpCell surface?
The UpCell surface is stable when standard cell culture conditions are applied, but scratching of the surface, for example, with a pipette tip, should be avoided, as this can impair the temperature-responsive properties of the product.
Can I reuse Nunc cultureware with UpCell surface?
Nunc cultureware with UpCell Surface are intended for single-use only. Cell-secreted and medium-derived components may adsorb to the surface during cell culture and compromise performance with repeated use.
How should I store Nunc cultureware with UpCell surface?
Nunc cultureware with UpCell Surface should be stored at room temperature (20-25 degrees C) away from direct sunlight.
What is the Nunc UpCell surface?
The Nunc UpCell surface comprises a temperature-responsive polymer that is covalently immobilized to the polystyrene surface of the culture area. The polymer becomes slightly hydrophobic above 32 degrees C, allowing cells to adhere. Below 32 degrees C, the polymer becomes hydrophilic, resulting in the binding of water to the surface and swelling of the surface, prompting cell detachment from the surface.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. For research and single-use only