Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 6 Well Plate, Nunclon Vita Surface, Pack of 1

Catalog No. 12567099 Shop All Thermo Scientific Products
Change view
Click to view available options
No. of Wells:
6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells
12-567-099 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12-567-099 Supplier Thermo Scientific™ Supplier No. 145380

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Grow temperamental cells directly on the Thermo Scientific™ Nunc™ Nunclon™ Vita 6-Well Multidish without matrix coatings or feeder layers.

Grow temperamental cells directly on the Thermo Scientific™ Nunc™ Nunclon™ Vita 6-Well Multidish without matrix coatings or feeder layers. The Nunclon Vita features a unique energy-treated polystyrene surface. When combined with conditioned media supplemented with a ROCK inhibitor, this surface supports attachment and expansion of human pluripotent stem cells.

Features:

  • Animal component-free surface
  • Consistent lot-to-lot performance
  • Supports attachment and growth of temperamental cells (e.g. HEK 293, MSC)
  • Supports attachment and expansion of human pluripotent stem cells in conditioned media containing Y27632 ROCK inhibitor
  • Allows non-enzymatic dissociation of human pluripotent stem cells with removal of ROCK inhibitor
TRUSTED_SUSTAINABILITY

Specifications

Description 6 well, Nunclon Vita
Culture Area 9.6 sq. cm
Material Polystyrene
Volume (Metric) Working 3 mL
Type Multiwell Plate
Quantity 1/Pk., 4/Cs.
Color Clear
Surface Treatment Nunclon Vita
Sterility Sterile
No. of Wells 6
Length (Metric) 128 mm
Width (Metric) 86 mm
Gridded Not gridded
Well Shape Flat Bottom (F-well)
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.