Learn More
Description
Perform molecular biology applications with non-treated Thermo Scientific™ Nunc™ Rectangular Dishes, which have ANSI™ standard dimensions and are optimized for robotics.
Rectangular Dishes 1, 4 and 8-Well
- 1-well dish is ideal for HTS Petri dish applications
- 4- and 8-well dishes are ideal for assay compartmentalization, probe multiple gene sub-set
- 4-well dish holds four glass slides for storage
1-Well Format
- Convenient plate format for cell culture
- HTS Petri dish applications
4- and 8-Well Formats
- Assay compartmentalization
- Probe multiple gene subsets
- Wash Western blot strips
- 4-well dish holds four glass slides
- Useful in all areas of cell culture, including scale-up and cloning
Ordering Information:
Customized bar coding is available
Specifications
Specifications
| Barcode | No Barcode |
| Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
| Format | SBS Standard |
| Array | 1 x 4 |
| Color | Clear |
| Product Type | Cell Culture Dish |
| Material | Polystyrene |
| No. of Wells | 4 |
| Sterility | Sterile |
| Surface Treatment | Non-treated |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.