Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ Rectangular Dishes

Catalog No. 12565497 Shop All Thermo Scientific Products
Change view
Click to view available options
No. of Wells:
1
4
8
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Wells
12-565-497 8
12-565-493 1
12-565-495 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 12-565-497 Supplier Thermo Scientific™ Supplier No. 267062
  • $1,085.86
    $787.65 / Case of 100

    Save $298.21

    27% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Perform molecular biology applications with non-treated Thermo Scientific™ Nunc™ Rectangular Dishes, which have ANSI™ standard dimensions and are optimized for robotics.

Rectangular Dishes 1, 4 and 8-Well

  • 1-well dish is ideal for HTS Petri dish applications
  • 4- and 8-well dishes are ideal for assay compartmentalization, probe multiple gene sub-set
  • 4-well dish holds four glass slides for storage

1-Well Format

  • Convenient plate format for cell culture
  • HTS Petri dish applications

4- and 8-Well Formats

  • Assay compartmentalization
  • Probe multiple gene subsets
  • Wash Western blot strips
  • 4-well dish holds four glass slides
  • Useful in all areas of cell culture, including scale-up and cloning

Ordering Information:

Customized bar coding is available

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Format SBS Standard
Array 2 x 4
Color Clear
Product Type Cell Culture Dish
Material Polystyrene
No. of Wells 8
Sterility Sterile
Surface Treatment Non-treated
Total Volume (Metric) Well 11 mL
Well Shape Rectangular
Includes Lid
No. per Case 100/Cs.
No. per Pack 10 Pk.
Quantity 10 Pk.
Length (Metric) 100 mm
Width (Metric) 86 mm
Height (Metric) 59 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.