Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ Square BioAssay Dishes

Catalog No. p-4524665 Shop All Thermo Scientific Products
Change view
Click to view available options
Height (Metric):
20 mm
25 mm
Surface Treatment:
Nunclon Delta
Untreated
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (Metric) Surface Treatment
12-565-101 25 mm Nunclon Delta
12-565-224 25 mm Untreated
12-565-743 20 mm Untreated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 12-565-101 Supplier Thermo Scientific™ Supplier No. 166508
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Screen large numbers of colonies for cloning experiments, culturing bacteria, fungi and agar diffusion assays using Thermo Scientific™ Nunc™ Squre BioAssay dishes.

Available in cell culture-treated and non-treated surfaces, the versatile Thermo Scientific™ Nunc™ Square BioAssay dishes are useful for screening large numbers of colonies, culturing bacteria and fungi, or providing a contained environment for other culture vessels.

  • Use as a moisture chamber for incubation on Nunc MicroWell™ Plates
  • Lined with filter paper it is ideal as an incubation chamber for in situ hybridization
  • Optically clear and compatible with automated robotic instrumentation

Untreated Dishes

  • Ideal for screening libraries or isolating clones
  • Useful for screening large numbers of colonies in cloning experiments
  • Can be used for culturing of bacteria and fungi as well as a moisture chamber for incubation of Nunc MicroWell Plates
  • Suitable in agar diffusion assay
  • Lined with wet filter paper, it is also useful as an incubation chamber for in situ hybridization
  • Large growth area
  • Designed for genomic screen and colony picking
TRUSTED_SUSTAINABILITY

Specifications

Bottom Shape Flat
Capacity (Metric) 135 mL
Color Clear
Culture Area 500 sq. cm
Shape Square
Sterility Sterile
Surface Treatment Nunclon Delta
Type Multi-Well Plate
Volume (Metric) Working 135 mL
No. per Case 16
No. per Pack 4
Height (Metric) 25 mm
Height (English) 25 in.
Show More Show Less
What is the difference between the Nunclon Delta surface and a tissue culture (TC) surface from a competitor?

Our Nunclon Delta surface for adherent cells is a standard tissue culture (TC) surface modification that makes the polystyrene surface more hydrophilic. This facilitates maximum adhesion for a broad range of cell types.

The Nunclon Delta surface is Thermo Scientific Nunclon Delta-certified, which is a Thermo Fisher proprietary certification that has a quality standard to help ensure consistency. Every lot of Thermo Scientific Nunclon Delta-certified cell cultureware is tested with Gibco media for:
- Less than 10% growth variation in L929, Hel299 and primary chick embryo cells
- Less than 15% variation colony formation of V79-4 cells

More information and all applicable products can be found in the "Product selection guide for every step of your cell culture workflow" brochure .

Do you offer a Nunc square petri dish with a grid on the bottom?

We offer Nunc Square BioAssay Dishes, but this product does not have a grid.

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.