Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ Storage Vials with Caps
GREENER_CHOICE

Catalog No. 0974071A
Change view
Click to view available options
Volume (Metric) Working:
0.5 mL
2.0 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Working
09-740-71A 0.5 mL
09-740-71B 2.0 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-740-71A Supplier Thermo Scientific™ Supplier No. 264262
Add to Cart
Add to Cart

Assure safe centrifugation and storage of PCR reagents, restriction enzymes, and other diagnostic and biochemical reagents or samples by using these storage vials with caps.

Assure safe centrifugation and storage of PCR reagents, restriction enzymes and other biochemical reagents and samples by using Thermo Scientific™ Nunc™ Storage Vials with Caps. Available in 0.5mL and 2.0mL sizes with frosted writing area, the vials have excellent chemical resistance with leakproof closure.

Features:

  • Made from high-purity polypropylene copolymer with excellent chemical resistance
  • Leakproof high-density polyethylene screw closure
  • Centrifuged to a maximum of 13,000 x g
  • Temperature range: -40°C to 95°C One full turn removes the ergonomic closure
  • RNase/DNase-free

Ordering Information:

Racks 264263 and 270972 are compatible with these vials. Color coders available: 254416, 264335, 264408, 264440, 264459, 264467, 264491.

TRUSTED_SUSTAINABILITY

Specifications

Material PPCO
Sterility Sterile
Volume (Metric) 0.9 mL
No. per Case 500
No. per Pack 500
Volume (Metric) Working 0.5 mL
Product Line Nunc
Volume (Metric) Total 0.9 mL
Closure Material Polyethylene
Closure Type Screw Cap
Quantity 500 Cs.
For Use With (Equipment) 264263, 270972 Rack
Certifications/Compliance Certificate of Analysis (COA), Certificate of Conformance (COC)
Product Type Storage Vial with Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.