Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nunc™ Universal and Transport Containers with Caps

Catalog No. p-4525877
Change view
Click to view available options
Material:
Polystyrene
Sterility:
Non-sterile
Sterile
Label:
Labeled
No Label
Includes:
No Spoon
Spoon
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Sterility Label Includes
12-565-205 Polystyrene Non-sterile Labeled No Spoon
12-565-206 Polystyrene Sterile No Label No Spoon
12-565-209 Polystyrene Sterile Labeled No Spoon
12-565-208 Polystyrene Sterile Labeled Spoon
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12-565-205 Supplier Thermo Scientific™ Supplier No. 363282
Only null left
Add to Cart
Add to Cart

Store and transport blood, urine, fecal and other specimens safely using these universal and transport containers with caps.

Safely store and transport blood, urine, fecal and other specimens using Thermo Scientific™ Nunc™ Universal and Transport Containers with Caps. These containers are designed with inner and outer containers of high mechanical strength. Caps are designed to avoid droplets adhering to the surface, reducing risk of aerosols. Nunc Labware Products are made from high purity resins, and molded using our state-of-the-art processes. Plastic labware is a safer alternative to glass without sacrificing accuracy.

Features:

Universal Containers

  • Working volume: 25mL
  • Outer case available in transparent or opaque material
  • Made of polystyrene with polyethylene caps (except 374799 which has polystyrene body with polyethylene, high-impact polystyrene and low density polystyrene)
  • Available sterile or nonsterile, and with or without a spoon for handling of solid and semi-solid samples
  • Some available with writing area
  • Max. RCF tested: 4000

Transport Container

  • Polypropylene with polyethylene cap or high-impact polystyrene with polyethylene
  • 374799 is comprised of a universal container with writing area and spoon in a compact transport container
TRUSTED_SUSTAINABILITY

Specifications

Product Type Universal Container
Closure Included Yes
Material Polystyrene
Sterility Non-sterile
Capacity (Metric) 25 mL
Certifications/Compliance Transport and Universal Containers have passed the 95 kPa test described in UN packaging instructions 602/650 for transport of infectious substances and diagnostic specimens.
Closure Material Polyethylene
Color Clear
Description W/o spoon; Nonsterile; PS; Writing area
Includes No Spoon
Label Labeled
No. per Case 300/Cs.
No. per Pack 50/Pk.
RCF Rating 4000 x g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.