Learn More
Cayman Chemical N-undecanoyl-L-Homoserine lactone, Purity: ≥98%, CAS: 216596-71-3, 50 mg
SDP
Catalog No. 502966538
Encompass_Preferred
A bacterial quorum-sensing signaling molecule; induces GFP production in E. coli expressing the V. fischeri LuxR-PluxI quorum-sensing operon from 100-1,000 nM; inhibits concanavalin A-induced proliferation of isolated mouse splenocytes (IC50 = 90 µM)
Catalog No. 50-296-6538 Supplier Cayman Chemical Supplier No. 1682750 MG
A bacterial quorum-sensing signaling molecule; induces GFP production in E. coli expressing the V. fischeri LuxR-PluxI quorum-sensing operon from 100-1,000 nM; inhibits concanavalin A-induced proliferation of isolated mouse splenocytes (IC50 = 90 µM)