Learn More
ABclonal Technology Nup107 Rabbit mAb
SDP
Catalog No. 5000008078
Encompass_Preferred
This gene encodes a member of the nucleoporin family The protein is localized to the nuclear rim and is an essential component of the nuclear pore complex (NPC) All molecules entering or leaving the nucleus either diffuse through or are actively transported by the NPC Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized
Catalog No. 50-000-08078 Supplier ABclonal Technology Supplier No. A2095520UL
This gene encodes a member of the nucleoporin family The protein is localized to the nuclear rim and is an essential component of the nuclear pore complex (NPC) All molecules entering or leaving the nucleus either diffuse through or are actively transported by the NPC Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized