Learn More
Aviva Systems Biology Corp NUP153 Antibody - C-terminal region (ARP61446_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61446_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NUP153. It was validated on Western Blot. . Alternate names: HNUP153, N153ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-287-381