Learn More
Aviva Systems Biology Corp NUP155 Antibody - N-terminal region (ARP53619_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53619_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NUP155. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human NUP155. Alternate names: KIAA0791, N155ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-858