Learn More
Aviva Systems Biology Corp Nup160 Antibody - C-terminal region (ARP61864_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61864_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Nup160. It was validated on Western Blot. . Alternate names: 160kDa, 2810011M03Rik, AA414952, AU020188, Gtl-13, Gtl1-13, Gtl13, KIAA0197, mKIAA0197ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-489