Learn More
Aviva Systems Biology Corp Nup37 Antibody - N-terminal region (ARP46327_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46327_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Nup37. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2410003L22Rik, 2810039M17RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-249