Learn More
Aviva Systems Biology Corp NUP50 Antibody - C-terminal region (ARP52288_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52288_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NUP50. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human NUP50. Alternate names: MGC39961, NPAP60, NPAP60LApplicationIF, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-60710