Learn More
Enzo Life Sciences Nupherin™ Transfection Reagent, (0.75mg)
SDP
Catalog No. 501031797
Encompass_Preferred
Greatly enhances transfection efficiency. Use/Stability: On first use, aliquot the reagent and store at -80°C. Thawed aliquots of Nupherin™ can be frozen after use for. Long Term Storage or refrigerated for up to 1 week. Do not dilute reagent. Long Term Storage: -80°C
Catalog No. 50-103-1797 Supplier Enzo Life Sciences Supplier No. BMLSE2250075
Greatly enhances transfection efficiency. Use/Stability: On first use, aliquot the reagent and store at -80°C. Thawed aliquots of Nupherin™ can be frozen after use for. Long Term Storage or refrigerated for up to 1 week. Do not dilute reagent. Long Term Storage: -80°C