Learn More
Grass Technologies NUPREP SKIN GEL(3 TUBE PK)
Catalog No. NC9005911
Encompass
NUPREP EEG Skin Prepping Gel, 4 oz. Tube, Pleasantly Formulated Skin Prep Gel lowers Impedance at Electrode-Skin Junction, Mildly Abrasive, 3/BX
Catalog No. NC9005911 Supplier Grass Technologies Supplier No. 122736100
NUPREP EEG Skin Prepping Gel, 4 oz. Tube, Pleasantly Formulated Skin Prep Gel lowers Impedance at Electrode-Skin Junction, Mildly Abrasive, 3/BX