Learn More
Medchemexpress LLC Nur77 modulator 3 | 2097787-75-0 | 99.0% | 308.40 | 100 MG
SDP
Catalog No. 5000391902
Encompass_Preferred
Nur77 modulator 3 is a Nur77 modulator that induces Nur77 expression, inhibits hepatic stellate cells (HSCs) activation, and reduces extracellular matrix (ECM) deposition. It enhances Nur77-dependent autophagic flux and significantly inhibits the mTORC1 signaling pathway. This modulator can ameliorate HSCs activation, inflammation, and hepatic fibrosis *in vivo*.
  • Induces Nur77 expression
  • Inhibits hepatic stellate cells (HSCs) activation
  • Reduces extracellular matrix (ECM) deposition
  • Enhances Nur77-dependent autophagic flux
  • Significantly inhibits the mTORC1 signaling pathway
  • Ameliorates HSCs activation, inflammation, and hepatic fibrosis *in vivo*
Catalog No. 50-003-91902 Supplier Medchemexpress LLC Supplier No. HY162147100MG
Nur77 modulator 3 is a Nur77 modulator that induces Nur77 expression, inhibits hepatic stellate cells (HSCs) activation, and reduces extracellular matrix (ECM) deposition. It enhances Nur77-dependent autophagic flux and significantly inhibits the mTORC1 signaling pathway. This modulator can ameliorate HSCs activation, inflammation, and hepatic fibrosis *in vivo*.
  • Induces Nur77 expression
  • Inhibits hepatic stellate cells (HSCs) activation
  • Reduces extracellular matrix (ECM) deposition
  • Enhances Nur77-dependent autophagic flux
  • Significantly inhibits the mTORC1 signaling pathway
  • Ameliorates HSCs activation, inflammation, and hepatic fibrosis *in vivo*