Learn More
Cayman Chemical nuregulIn1 TYP I2b EXra 100ug
SDP
Catalog No. 502948662
Encompass_Preferred
Source: Recombinant human C-terminal His-tagged neuregulin-1 type I-β2b extracellular domain expressed in HEK293 cells • Amino acids: 20-238 • MW: 25.3 kDa
Catalog No. 50-294-8662 Supplier Cayman Chemical Supplier No. 32094100 UG
Source: Recombinant human C-terminal His-tagged neuregulin-1 type I-β2b extracellular domain expressed in HEK293 cells • Amino acids: 20-238 • MW: 25.3 kDa