Learn More
Sklar Instruments Nurse's Bandage Scissors, Sklar 5-1/2 in., Angled, Smooth, with Pocket Clip, Finger Rings, Stainless Steel. EA.
SDP
Catalog No. 501184183 Shop All Sklar Instruments Products
Encompass_Preferred
Nurse's Bandage Scissors, Sklar. 5-1/2 in., Angled, Smooth, with Pocket Clip, Finger Rings, Stainless Steel. Surgi-OR Nurse's Bandage Scissors are non-ratcheted, finger ring scissors primarily used for removing tight bandages. The tip of the lower blade features a flattened blunt nodule that is intended to slide between bandages and skin without harming the skin. The shanks are straight and feature a pocket clip. After the screw lock the blades angle slightly. The blades of this product are smooth and both tips are blunt. The overall length of the instrument is 5-1/2 inches.
Catalog No. 50-118-4183 Supplier Sklar Instruments Supplier No. 95232
Nurse's Bandage Scissors, Sklar. 5-1/2 in., Angled, Smooth, with Pocket Clip, Finger Rings, Stainless Steel. Surgi-OR Nurse's Bandage Scissors are non-ratcheted, finger ring scissors primarily used for removing tight bandages. The tip of the lower blade features a flattened blunt nodule that is intended to slide between bandages and skin without harming the skin. The shanks are straight and feature a pocket clip. After the screw lock the blades angle slightly. The blades of this product are smooth and both tips are blunt. The overall length of the instrument is 5-1/2 inches.