Learn More
Aviva Systems Biology Corp NUSAP1 Antibody - middle region (ARP46285_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46285_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NUSAP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NUSAP1. Alternate names: ANKT, BM037, FLJ13421, LNP, PRO0310p1, Q0310, SAPL, NUSAPApplicationWB. Species reactivity: Human, Rabbit
Catalog No. 50-014-59294