Learn More
Aviva Systems Biology Corp NUSAP1 Antibody - C-terminal region (ARP46245_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46245_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NUSAP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human NUSAP1. Alternate names: ANKT, BM037, FLJ13421, LNP, PRO0310p1, Q0310, SAPL, NUSAPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59286