Learn More
Nusep Inc NUVIEW UNIV GEL 4-20 10-WELL
Catalog No. NC1854204
Encompass_Preferred
nUView Tris-Glycine SDS-PAGE Precast Gel 4-20% 10well. Cassette size: 10 x 8 cm. Fits Hoefer, Cytiva, Merck Tanks. 10 gel cassettes per box. All nUView™ precast gels incorporate a unique formulation allowing protein bands to be visualized in only 2 minutes in the presence of ultraviolet (UV) light. Simply place your gel over a 250-320 nm UV trans-illuminator and watch high-resolution bands appear in just 2 minutes. With nUView™ technology you no longer have to stain and de-stain your gels – increasing protein recovery and saving you significant time while receiving improved results.
Catalog No. NC1854204 Supplier Nusep Inc Supplier No. NG10420
nUView Tris-Glycine SDS-PAGE Precast Gel 4-20% 10well. Cassette size: 10 x 8 cm. Fits Hoefer, Cytiva, Merck Tanks. 10 gel cassettes per box. All nUView™ precast gels incorporate a unique formulation allowing protein bands to be visualized in only 2 minutes in the presence of ultraviolet (UV) light. Simply place your gel over a 250-320 nm UV trans-illuminator and watch high-resolution bands appear in just 2 minutes. With nUView™ technology you no longer have to stain and de-stain your gels – increasing protein recovery and saving you significant time while receiving improved results.