Learn More
Nusep Inc NUSEP SNGLESHOT LB AGAR 20/PK
Catalog No. NC1854235
Encompass_Preferred
NuSep's SingleShot LB Agar is packaged in 20 pouches per box, providing convenience for microbiological applications. Each pre-measured pouch, when dissolved in 50 mL of water, yields a 1x working solution, simplifying the preparation process and ensuring consistent results.Specifications:-Contents: 20 individual pouches per box-Yield: Each pouch makes 50 mL of 1x working solution-Convenience: Pre-measured for quick and reliable preparation-Use: Ideal for bacterial cultivation in laboratory settingsNuSep's SingleShot LB Agar is designed to deliver dependable performance, making it a practical choice for labs looking for efficient and consistent agar preparation.
Catalog No. NC1854235 Supplier Nusep Inc Supplier No. DBMLBA01
NuSep's SingleShot LB Agar is packaged in 20 pouches per box, providing convenience for microbiological applications. Each pre-measured pouch, when dissolved in 50 mL of water, yields a 1x working solution, simplifying the preparation process and ensuring consistent results.Specifications:-Contents: 20 individual pouches per box-Yield: Each pouch makes 50 mL of 1x working solution-Convenience: Pre-measured for quick and reliable preparation-Use: Ideal for bacterial cultivation in laboratory settingsNuSep's SingleShot LB Agar is designed to deliver dependable performance, making it a practical choice for labs looking for efficient and consistent agar preparation.