Learn More
Nusep Inc NUSEP TGS BUFF 5L PDR SACHET
Catalog No. NC1854223
Encompass_Preferred
NuSep's SDS Tris-Glycine Sachet is designed for easy preparation of electrophoresis buffer. When dissolved, each sachet produces 5,000 mL of pH 8.5 Tris-Glycine-SDS buffer, ideal for use with Tris-Glycine electrophoresis gels.Specifications:-pH Value: 8.5-Working Volume: 5,000 mL per sachet-Storage: Store at room temperature for short-term use or at 4°C for long-term storageThis buffer offers reliable performance for electrophoresis applications, ensuring consistent results with minimal preparation time. Simplify your lab work with NuSep's SDS Tris-Glycine sachet, providing convenience and dependable quality for your research needs.
Catalog No. NC1854223 Supplier Nusep Inc Supplier No. BG143
NuSep's SDS Tris-Glycine Sachet is designed for easy preparation of electrophoresis buffer. When dissolved, each sachet produces 5,000 mL of pH 8.5 Tris-Glycine-SDS buffer, ideal for use with Tris-Glycine electrophoresis gels.Specifications:-pH Value: 8.5-Working Volume: 5,000 mL per sachet-Storage: Store at room temperature for short-term use or at 4°C for long-term storageThis buffer offers reliable performance for electrophoresis applications, ensuring consistent results with minimal preparation time. Simplify your lab work with NuSep's SDS Tris-Glycine sachet, providing convenience and dependable quality for your research needs.