Learn More
Medchemexpress LLC Nusinersen | 1258984-36-9 | 7127.30 | 10 MG
SDP
Catalog No. 5000439225
Encompass_Preferred
Nusinersen is an antisense oligonucleotide active molecule that functions by modifying the pre-messenger RNA splicing of the SMN2 gene, promoting the production of full-length SMN protein. It has been shown to improve spinal muscular atrophy, ameliorating motor function and preventing motoneuron Cajal body disassembly in SMA mouse models.
  • Functions as an antisense oligonucleotide active molecule
  • Modifies pre-messenger RNA splicing of the SMN2 gene
  • Promotes production of full-length SMN protein
  • Improves spinal muscular atrophy
  • Ameliorates motor function in SMA mouse models
  • Prevents motoneuron Cajal body disassembly
  • Rescues skeletal muscle fiber size and oxidative metabolism
Catalog No. 50-004-39225 Supplier Medchemexpress LLC Supplier No. HY11298010MG
Nusinersen is an antisense oligonucleotide active molecule that functions by modifying the pre-messenger RNA splicing of the SMN2 gene, promoting the production of full-length SMN protein. It has been shown to improve spinal muscular atrophy, ameliorating motor function and preventing motoneuron Cajal body disassembly in SMA mouse models.
  • Functions as an antisense oligonucleotide active molecule
  • Modifies pre-messenger RNA splicing of the SMN2 gene
  • Promotes production of full-length SMN protein
  • Improves spinal muscular atrophy
  • Ameliorates motor function in SMA mouse models
  • Prevents motoneuron Cajal body disassembly
  • Rescues skeletal muscle fiber size and oxidative metabolism