Learn More
Medchemexpress LLC Nusinersen | 1258984-36-9 | 98.9% | 7127.3 g/mol | C234H340N61O128P17S17 | 5 MG
SDP
Catalog No. 5000262983
Encompass_Preferred
Nusinersen is an antisense oligonucleotide that modulates SMN2 pre-mRNA splicing to increase production of full-length survival motor neuron (SMN) protein. This research-grade material (CAS 1258984-36-9) is supplied as a purified oligonucleotide for laboratory biochemical and cellular studies; typical pack size is 5 mg and reported purity is 98.91%.
  • Antisense oligonucleotide for SMN2 splicing modulation.
  • High purity (approximately 98.9%) suitable for biochemical assays.
  • Supplied as a solid oligonucleotide for in vitro and ex vivo use.
  • Applicable to studies of spinal muscular atrophy mechanisms and SMN biology.
  • Characterized by defined molecular formula and molecular weight for reproducible experiments.
Catalog No. 50-002-62983 Supplier Medchemexpress LLC Supplier No. HY1129805MG
Nusinersen is an antisense oligonucleotide that modulates SMN2 pre-mRNA splicing to increase production of full-length survival motor neuron (SMN) protein. This research-grade material (CAS 1258984-36-9) is supplied as a purified oligonucleotide for laboratory biochemical and cellular studies; typical pack size is 5 mg and reported purity is 98.91%.
  • Antisense oligonucleotide for SMN2 splicing modulation.
  • High purity (approximately 98.9%) suitable for biochemical assays.
  • Supplied as a solid oligonucleotide for in vitro and ex vivo use.
  • Applicable to studies of spinal muscular atrophy mechanisms and SMN biology.
  • Characterized by defined molecular formula and molecular weight for reproducible experiments.