Learn More
Trajan Scientific VICI Valco Nut, Internal Nut - High Pressure, PEEK, 1/16" Fingertight, 0.88" Length, used in Cheminert Valves With ZDV Fittings, 10 Pack
Catalog No. 501090228
Encompass
VICI Valco Nut, Internal Nut - High Pressure, PEEK, 1/16" Fingertight, 0.88" Length, used in Cheminert Valves With ZDV Fittings. Made in the USA.
Catalog No. 50-109-0228 Supplier Trajan Scientific Supplier No. ZN1FPK10
VICI Valco Nut, Internal Nut - High Pressure, PEEK, 1/16" Fingertight, 0.88" Length, used in Cheminert Valves With ZDV Fittings. Made in the USA.