Learn More
Phenomenex Inc Nut and Ferrule Set, S.S.
Catalog No. 502399677 Shop All Phenomenex Products
Encompass_Preferred
Nut and Ferrule Set, S.S. for 1/16in OD Tubing, 10-32 Threads
Catalog No. 50-239-9677 Supplier Phenomenex Inc Supplier No. AQ03018
Nut and Ferrule Set, S.S. for 1/16in OD Tubing, 10-32 Threads