Learn More
IDEX Health & Science Upchurch NUT FEMALE PEEK MCR TEE/CROSS
Catalog No. NC1919729
Encompass_Preferred
Nut, Fingertight; Idex; MicroTight; Female nut; Internal threads: 5/16-24; Color: natural; Material: PEEK
Catalog No. NC1919729 Supplier IDEX Health & Science Upchurch Supplier No. P416
Nut, Fingertight; Idex; MicroTight; Female nut; Internal threads: 5/16-24; Color: natural; Material: PEEK