Learn More
IDEX Health & Science Upchurch NUT FLANGLESS GREEN 1/8IN 10PK
Catalog No. NC1919051
Encompass_Preferred
Idex Flangeless Nuts: Compatible with 1/4-28, Delrin, Green, For Use With: For 0.13 in. O.D. Tubing, Max. Pressure: 500 psi, Flangeless standard knurled head nut, Certifications/Compliance: ROHS, Port Style: 0.25 to 28 flat bottom, Quantity: 10/Pk.
Catalog No. NC1919051 Supplier IDEX Health & Science Upchurch Supplier No. P305X
Idex Flangeless Nuts: Compatible with 1/4-28, Delrin, Green, For Use With: For 0.13 in. O.D. Tubing, Max. Pressure: 500 psi, Flangeless standard knurled head nut, Certifications/Compliance: ROHS, Port Style: 0.25 to 28 flat bottom, Quantity: 10/Pk.