Learn More
IDEX Health & Science Upchurch NUT FLNGLSS RED 1/16IN 10PK
Catalog No. NC1919748
Encompass_Preferred
Idex Flangeless Nuts: Compatible with 1/4-28, Delrin, Red, For Use With: For 0.06 in. O.D. Tubing, Max. Pressure: 2000 psi, Flangeless standard knurled head nut, Certifications/Compliance: ROHS, Port Style: 0.25 to 28 flat bottom, Quantity: 10/Pk.
Catalog No. NC1919748 Supplier IDEX Health & Science Upchurch Supplier No. P202X
Idex Flangeless Nuts: Compatible with 1/4-28, Delrin, Red, For Use With: For 0.06 in. O.D. Tubing, Max. Pressure: 2000 psi, Flangeless standard knurled head nut, Certifications/Compliance: ROHS, Port Style: 0.25 to 28 flat bottom, Quantity: 10/Pk.