Learn More
Perkin Elmer US LLC Injector Adapter Nut for Optima 2x00/4x00/5x00/7x00/8x00
Catalog No. 509043911
Encompass_Preferred
This nut is included in the Organics Sample Introduction Kit for Optima 8x00 (N0780608) and the Organics Sample Introduction Kit for Optima 2x00/4x00/5x00/7x00 DV (N0770420).
Catalog No. 50-904-3911 Supplier Perkin Elmer US LLC Supplier No. N0776025
This nut is included in the Organics Sample Introduction Kit for Optima 8x00 (N0780608) and the Organics Sample Introduction Kit for Optima 2x00/4x00/5x00/7x00 DV (N0770420).