Learn More
IDEX Health & Science Upchurch NUT LITETOUCH 1/16IN 10PK
Supplier:  IDEX Health & Science Upchurch LT110X
Encompass_Preferred
Idex LiteTouch Nut, PEEK, 1/16 in. OD Tubing, 10-32 Coned, Description: Fits 1/16 in. O.D. tubing, For Use With: LiteTouch Ferrule System, Thread Size: 10 to 32 UNF
Catalog No. NC1919413