Learn More
Cole-Parmer Idex Flangeless Male Nut, Standard Knurl, Natural PFA, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Catalog No. NC2101627 Shop All Cole-Parmer Products
Encompass
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.
This item is not returnable. View return policy
Catalog No. NC2101627 Supplier Cole-Parmer Supplier No. 0201976
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.