Learn More
Grainger Nut Runner L 11 in
Catalog No. 502765204
Encompass_Preferred
The Greenlee 16V991 is a nut runner designed for 1-1/2" and 1-1/8" conduit fittings. It features a rugged steel housing with rubber grip, reversible ratchet head that provides high torque, and a 1/2" square drive. This nut runner has a compact 10" length that allows access in tight spaces. It requires an impact wrench or drill for power. The 16V991 nut runner enables efficient tightening and loosening of conduit fittings up to 1-1/2" trade size.
Catalog No. 50-276-5204 Supplier Grainger Supplier No. 16V991
The Greenlee 16V991 is a nut runner designed for 1-1/2" and 1-1/8" conduit fittings. It features a rugged steel housing with rubber grip, reversible ratchet head that provides high torque, and a 1/2" square drive. This nut runner has a compact 10" length that allows access in tight spaces. It requires an impact wrench or drill for power. The 16V991 nut runner enables efficient tightening and loosening of conduit fittings up to 1-1/2" trade size.