Learn More
MSC Value Collection J Nut, Spring Steel, 1/4 in Screw Size
Catalog No. 502837312
Encompass
This J nut is designed for secure fastening applications, featuring a durable spring steel construction. It is compatible with 1/4 in screws and is suitable for material thicknesses ranging from 1/16 to 3/32 in, providing a reliable and efficient solution for assembly needs.
  • Constructed from durable spring steel.
  • Compatible with 1/4 in screw sizes.
  • Suitable for material thickness from 1/16 to 3/32 in.
  • Features a J nut type design.
  • Designed for inch system measurements.
  • Supplied in a pack of 50 units.
Catalog No. 50-283-7312 Supplier MSC Supplier No. 66434481
This J nut is designed for secure fastening applications, featuring a durable spring steel construction. It is compatible with 1/4 in screws and is suitable for material thicknesses ranging from 1/16 to 3/32 in, providing a reliable and efficient solution for assembly needs.
  • Constructed from durable spring steel.
  • Compatible with 1/4 in screw sizes.
  • Suitable for material thickness from 1/16 to 3/32 in.
  • Features a J nut type design.
  • Designed for inch system measurements.
  • Supplied in a pack of 50 units.