Learn More
IDEX Health & Science Upchurch NUT SUPR FLNGLS 1/8 DELRIN BK
Catalog No. NC1919707
Encompass_Preferred
Idex Flat-Bottom Fittings, Black, Diameter Outer: 0.125 in., 0.318 cm, Delrin (Acetal), Nut, Pressure: 2500 psi
Catalog No. NC1919707 Supplier IDEX Health & Science Upchurch Supplier No. P332
Idex Flat-Bottom Fittings, Black, Diameter Outer: 0.125 in., 0.318 cm, Delrin (Acetal), Nut, Pressure: 2500 psi