Learn More
IDEX Health & Science Upchurch NUT SUPR FLNGLSS PEEK 10/PK
Catalog No. NC1919750
Encompass_Preferred
Idex Male-Threaded Nuts, For Use With: 1/16 in. O.D. Tubing, Length: 0.87 in., PEEK, Width: 0.365 in., Natural, Diameter Outer: 0.0625 in.
Catalog No. NC1919750 Supplier IDEX Health & Science Upchurch Supplier No. P255X
Idex Male-Threaded Nuts, For Use With: 1/16 in. O.D. Tubing, Length: 0.87 in., PEEK, Width: 0.365 in., Natural, Diameter Outer: 0.0625 in.