Learn More
Perkin Elmer US LLC Nut - TBG Flanged 0.125 PEEK
Catalog No. 502117123
Encompass_Preferred
This PEEK nut is compatible with GemTip Cross-Flow II Nebulizers for use in Optima 2x00/4x00/5x00/7x00 DV series, Optima 8x00 and Avio 200/500 ICP-OES series instruments.
Catalog No. 50-211-7123 Supplier Perkin Elmer US LLC Supplier No. 09920545
This PEEK nut is compatible with GemTip Cross-Flow II Nebulizers for use in Optima 2x00/4x00/5x00/7x00 DV series, Optima 8x00 and Avio 200/500 ICP-OES series instruments.