Learn More
IDEX Health & Science Upchurch NUT UPTIGHT MALE 10/PK
Catalog No. NC1919185
Encompass_Preferred
Idex Male-Threaded Nuts, For Use With: 1/16 in. O.D. Tubing, Length: 0.4 in., Stainless Steel, Width: 0.25 in., Diameter Outer: 0.0625 in.
Catalog No. NC1919185 Supplier IDEX Health & Science Upchurch Supplier No. U400X
Idex Male-Threaded Nuts, For Use With: 1/16 in. O.D. Tubing, Length: 0.4 in., Stainless Steel, Width: 0.25 in., Diameter Outer: 0.0625 in.