Learn More
Medmark Technologies LLC NUTATOR MIXER 110 VOLT
Catalog No. NC0597936
Encompass_Preferred
NUTATOR MIXER 110 Volt. MedMark Lab Mixers. The original NUTATOR MIXER. A three dimensional rotating, rocking mixer at a 20 deg. angle @ 24rpms Ideal for hematology, blood-chemistry and other applications will hold up to 15 or more test tubes.
Catalog No. NC0597936 Supplier Medmark Technologies LLC Supplier No. NUTATOR110 V
NUTATOR MIXER 110 Volt. MedMark Lab Mixers. The original NUTATOR MIXER. A three dimensional rotating, rocking mixer at a 20 deg. angle @ 24rpms Ideal for hematology, blood-chemistry and other applications will hold up to 15 or more test tubes.