Learn More
Perkin Elmer US LLC FID Jet Replacement Tool
Catalog No. 509045112
Encompass_Preferred
0.25 in nut driver for removing jet from Auto-Ignite FID body.
Catalog No. 50-904-5112 Supplier Perkin Elmer US LLC Supplier No. N6101297
0.25 in nut driver for removing jet from Auto-Ignite FID body.