Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AlphaProTech NuTech™ Coverall with Elastic Wrist and Ankle and Serged Seams
BIOPHARMA

Catalog No. 19810568 Shop All AlphaProTech Products
Change view
Click to view available options
Size:
2X-Large/3X-Large
4X-Large/5X-Large
Large/X-Large
Small/Medium
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-810-568 4X-Large/5X-Large
19-810-565 Small/Medium
19-810-566 Large/X-Large
19-810-567 2X-Large/3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-810-568 Supplier AlphaProTech Supplier No. CV640228
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Strong, lightweight innovative microporous composite material protects the wearer from grime, dirt, and particulates.

  • Protects from grime, dirt and particulates
  • Pocketless to help control items entering and leaving work area
  • Tapered collar neckline and a front zipper for easy donning and doffing
  • With elastic wrist and ankle, serged seams

Specifications

Product Type Cleanroom Coveralls
Size 4X-Large/5X-Large
Material Microporous Composite Fabric
Color White
Disposable Single-use
Ankle Style Elastic
Neck Style Collared
Seam Type Serged
Sterility Non-sterile
For Use With (Application) Controlled Environments
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.