Learn More
AlphaProTech Critical Cover™ NuTech™ Frocks
BIOPHARMA
Catalog No. 19810569 Shop All AlphaProTech Products
Change view
Click to view available options
Size:
2X-Large/3X-Large
4X-Large/5X-Large
Large/X-Large
Small/Medium
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-810-569 Small/Medium
19-810-570 Large/X-Large
19-810-571 2X-Large/3X-Large
19-810-572 4X-Large/5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-810-569 Supplier AlphaProTech Supplier No. FK621212
Add to Cart
Add to Cart
This item is not returnable. View return policy
Microporous composite material frock is ideal for visitors and other wearers who do not require highly protective garments.
  • Protects clothes while offering comfort and breathability
  • Elastic wrist allows easy donning of gloves
  • Protective sleeves cut to an appropriate length as safeguard for full coverage
  • Snap collar provides additional coverage of the chest and neck area
Specifications
Size Small/Medium
Material Microporous Fabric
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Snaps
Cuff Style Elastic
Collar Type Snap
Sleeve Style Raglan Sleeve
Product Type Controlled Environments Frock
Seam Type Serged