Learn More
AlphaProTech NuTech™ Lab Coats
BIOPHARMA
Catalog No. 19810574 Shop All AlphaProTech Products
Change view
Click to view available options
Size:
2X-Large/3X-Large
4X-Large/5X-Large
Large/X-Large
Small/Medium
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-810-574 Large/X-Large
19-810-573 Small/Medium
19-810-575 2X-Large/3X-Large
19-810-576 4X-Large/5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-810-574 Supplier AlphaProTech Supplier No. LC626214
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Lightweight and breathable microporous composite material offers comfort and protection for a broad range of non-hazardous applications.
  • Strong breathable material offering protection
  • Non-confining tapered collar designed like shirt collar for style and comfort
  • Five-snap closure to provide fill coverage without gaps
  • Length approximately adjusted to size to provide additional coverage
  • Features tapered collar, elastic wrists, 3 pockets, snap close and serged seams
Specifications
Product Type Controlled Environments Lab Coat
Size Large/X-Large
Material Microporous Composite Fabric
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
No. of Pockets 3
Collar Type Tapered
Seam Type Serged