Learn More
Medchemexpress LLC HY-50696 10mg Medchemexpress, Nutlin-3 CAS:548472-68-0 Purity:>98%
SDP
Catalog No. 502029328
Encompass_Preferred
Medchemexpress, HY-50696 10mg Nutlin-3 CAS:548472-68-0 Nutlin-3 is a commercial available p53-MDM2 inhibitor, with Ki of 90 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9328 Supplier Medchemexpress LLC Supplier No. HY5069610MG
Medchemexpress, HY-50696 10mg Nutlin-3 CAS:548472-68-0 Nutlin-3 is a commercial available p53-MDM2 inhibitor, with Ki of 90 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.