Learn More
Cayman Chemical (±)-Nutlin-3, 548472-68-0, 10 mg
SDP
Supplier:  Cayman Chemical 1000437210
Encompass_Preferred
An inhibitor of p53-Mdm2 interaction (IC50 = 0.09 µM); induces the expression of p53-regulated genes and exhibits potent antiproliferative activity in cells with functional p53
Catalog No. NC9728454