Learn More
Enzo Life Sciences Nutlin-3 (25mg). CAS: 548472-68-0
SDP
Supplier:  Enzo Life Sciences ALX430128M025
Encompass_Preferred
(+/-) -Nutlin-3 is an inhibitor of p53/MDM2 interaction (IC50=0.09µM). Alternative name: (+/-)-4-[4,5-bis-(4-Chlorophenyl)-2-(2-isopropoxy-4-methoxyphenyl)-4,5-dihydroimidazole-1-carbonyl]-piperazin-2-one. Purity: ≥98% (HPLC (UV)). Appearance: White to off-white crystalline solid. Solubility: Soluble in methanol, 100% ethanol, DMSO, dimethyl formamide or acetonitrile; sparingly soluble in a 1:10 solution of ethanol:PBS, pH 7.2. Long Term Storage: -20°C.
Catalog No. 50-200-9218